Sodium methanethiosulphonate 97%

Synonyme: Sodium methanesulphonothioate

BD151527-100G BD151527-250MG BD151527-25G BD151527-5G BD151527-1G
BLDPBD151527-100GEA 1298 EUR
BLDPBD151527-100G BLDPBD151527-250MG BLDPBD151527-25G BLDPBD151527-5G BLDPBD151527-1G
Sodium methanethiosulphonate 97%
Sodium methanethiosulphonate

Storage condition: Inert atmosphere, Room Temperature

Formel: CH₃NaO₂S₂
Molecular Weight: 134,153 g/mol
Lagertemperatur: Ambient
MDL: MFCD03425630
CAS-Nummer: 1950-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR