Sodium-D-pantothenate 95%

Synonyme: Sodium (R)-3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoate,D-Pantothenic Acid Sodium Salt

BD151448-10G BD151448-25G BD151448-5G BD151448-100G
BLDPBD151448-10GEA 11 EUR
BLDPBD151448-10G BLDPBD151448-25G BLDPBD151448-5G BLDPBD151448-100G
Sodium-D-pantothenate 95%
Sodium-D-pantothenate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₁₇NO₅
Molecular Weight: 241,22 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00002767
CAS-Nummer: 867-81-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR