Natriumcyanat 97%

Synonyme: Cyansäure Natriumsalz

BD151414-100G BD151414-1000G BD151414-25G BD151414-500G
BLDPBD151414-100GEA 13 EUR
BLDPBD151414-100G BLDPBD151414-1000G BLDPBD151414-25G BLDPBD151414-500G
Natriumcyanat 97%
Natriumcyanat

Storage condition: Inert atmosphere, Room Temperature

Formel: CNNaO
Molecular Weight: 65,01 g/mol
Lagertemperatur: Ambient
MDL: MFCD00011121
CAS-Nummer: 917-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR