Benzo[h]chinolin 98%

Synonyme: 7,8-Benzoquinoline,alpha-Naphthoquinoline,4-Azaphenanthrene

BD15128-5G BD15128-25G BD15128-250MG BD15128-10G BD15128-1G BD15128-100G
BLDPBD15128-5GEA 45 EUR
BLDPBD15128-5G BLDPBD15128-25G BLDPBD15128-250MG BLDPBD15128-10G BLDPBD15128-1G BLDPBD15128-100G
Benzo[h]chinolin 98%
Benzo[h]chinolin

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₉N
Molecular Weight: 179,22 g/mol
Lagertemperatur: Ambient
MDL: MFCD00004984
CAS-Nummer: 230-27-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR