2-Brombenzophenon 95%

Synonyme: (2-Bromophenyl)(phenyl)methanone

BD15086-100G BD15086-5G BD15086-1G BD15086-25G BD15086-10G BD15086-250MG
BLDPBD15086-100GEA 514 EUR
BLDPBD15086-100G BLDPBD15086-5G BLDPBD15086-1G BLDPBD15086-25G BLDPBD15086-10G BLDPBD15086-250MG
2-Brombenzophenon 95%
2-Brombenzophenon

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₉BrO
Molecular Weight: 261,12 g/mol
Lagertemperatur: Ambient
MDL: MFCD00032462
CAS-Nummer: 13047-06-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR