(2-Amino-5-bromophenyl)(2-chlorophenyl)methanone 98%

BD15083-1G BD15083-5G
BLDPBD15083-1GEA 27 EUR
BLDPBD15083-1G BLDPBD15083-5G
(2-Amino-5-bromophenyl)(2-chlorophenyl)methanone 98%
(2-Amino-5-bromophenyl)(2-chlorophenyl)methanone

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₃H₉BrClNO
Molecular Weight: 310,57 g/mol
Lagertemperatur: Ambient
MDL: MFCD00156812
CAS-Nummer: 60773-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR