Salicylohydroxamsäure 97%

Synonyme: N,2-Dihydroxybenzamide,Salicylhydroxamic acid

BD150765-1G BD150765-5G BD150765-10G BD150765-500G BD150765-25G BD150765-100G
BLDPBD150765-1GEA 6 EUR
BLDPBD150765-1G BLDPBD150765-5G BLDPBD150765-10G BLDPBD150765-500G BLDPBD150765-25G BLDPBD150765-100G
Salicylohydroxamsäure 97%
Salicylohydroxamsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
Lagertemperatur: Ambient
MDL: MFCD00002110
CAS-Nummer: 89-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR