Salicylaldoxim 95%

BLDPBD150762-5GEA 7 EUR
BLDPBD150762-5G BLDPBD150762-0.5G BLDPBD150762-25G BLDPBD150762-500G BLDPBD150762-100G
Salicylaldoxim 95%
Salicylaldoxim

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₇NO₂
Molecular Weight: 137,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002120
CAS-Nummer: 94-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR