Safinamide 98%

Synonyme: (S)-2-[[4-[(3-Fluorobenzyl)oxy]benzyl]amino]propionamide,(S)-2-((4-((3-Fluorobenzyl)oxy)benzyl)amino)propanamide

BD150732-250MG BD150732-100MG
BLDPBD150732-250MGEA 16 EUR
BLDPBD150732-250MG BLDPBD150732-100MG
Safinamide 98%
Safinamide

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₇H₁₉FN₂O₂
Molecular Weight: 302,35 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00897036
CAS-Nummer: 133865-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR