S-Acetyl-L-glutathione 95%

Synonyme: N5-((R)-3-(Acetylthio)-1-((carboxymethyl)amino)-1-oxopropan-2-yl)-L-glutamine

BD150717-10G BD150717-250MG BD150717-25G BD150717-1G BD150717-5G
BLDPBD150717-10GEA 82 EUR
BLDPBD150717-10G BLDPBD150717-250MG BLDPBD150717-25G BLDPBD150717-1G BLDPBD150717-5G
S-Acetyl-L-glutathione 95%
S-Acetyl-L-glutathione

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₂H₁₉N₃O₇S
Molecular Weight: 349,36 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09952588
CAS-Nummer: 3054-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR