5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulphonate 97%

BLDPBD150680-250MGEA 25 EUR
BLDPBD150680-250MG BLDPBD150680-25G BLDPBD150680-5G BLDPBD150680-1G
5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulphonate 97%
5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulphonate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₄H₈F₆O₃S₂
Molecular Weight: 402,325 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236132
CAS-Nummer: 129946-88-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR