ɑ-(Trichloromethyl)benzyl acetate 98%

Synonyme: 2,2,2-Trichloro-1-phenylethyl acetate,Acetic Acid ɑ-(Trichloromethyl)benzyl Ester,Acetic Acid 2,2,2-Trichloro-1-phenylethyl Ester,2,2,2-Trichloro-1-phenylethyl Acetate

BD150534-25G BD150534-100G BD150534-500G
BLDPBD150534-25GEA 9 EUR
BLDPBD150534-25G BLDPBD150534-100G BLDPBD150534-500G
ɑ-(Trichloromethyl)benzyl acetate 98%
alpha-(Trichloromethyl)benzyl acetate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₉Cl₃O₂
Molecular Weight: 267,53 g/mol
Lagertemperatur: Ambient
MDL: MFCD00000804
CAS-Nummer: 90-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR