Sodium hydroxymethanesulphinate 95%

Synonyme: Sodium formaldehydesulphoxylate

BD150510-100G
BLDPBD150510-100GEA 7 EUR
BLDPBD150510-100G
Sodium hydroxymethanesulphinate 95%
Sodium hydroxymethanesulphinate
Formel: CH₃NaO₃S
Molecular Weight: 118,09 g/mol
MDL: MFCD00040426
CAS-Nummer: 149-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR