Bisindolylmaleimide IX methanesulphonate 98%

BLDPBD150474-25MGEA 81 EUR
BLDPBD150474-25MG BLDPBD150474-50MG
Bisindolylmaleimide IX methanesulphonate 98%
Bisindolylmaleimide IX methanesulphonate

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₂₅H₂₃N₅O₂S·CH₄O₃S
Molecular Weight: 457,6·96,1 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD03788207
CAS-Nummer: 138489-18-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR