Resorcinol-O,O'-diacetic acid 98%

Synonyme: 2,2'-(1,3-Phenylenebis(oxy))diacetic acid

BD150311-5G BD150311-1G BD150311-25G
BLDPBD150311-5GEA 135 EUR
BLDPBD150311-5G BLDPBD150311-1G BLDPBD150311-25G
Resorcinol-O,O'-diacetic acid 98%
Resorcinol-O,O'-diacetic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₀O₆
Molecular Weight: 226,19 g/mol
Lagertemperatur: Ambient
MDL: MFCD00016696
CAS-Nummer: 102-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR