2-Chlorthioxanthon 98%

Synonyme: 2-Chlorothioxanthen-9-one,2-Chloro-9H-thioxanthen-9-one

BD15021-5G BD15021-10G BD15021-25G
BLDPBD15021-5GEA 7 EUR
BLDPBD15021-5G BLDPBD15021-10G BLDPBD15021-25G
2-Chlorthioxanthon 98%
2-Chlorthioxanthon

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₃H₇ClOS
Molecular Weight: 246,72 g/mol
Lagertemperatur: Ambient
MDL: MFCD00005067
CAS-Nummer: 86-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR