(R)-3-Phenylpiperidine 97%

BLDPBD150175-100MGEA 145 EUR
BLDPBD150175-100MG BLDPBD150175-250MG BLDPBD150175-1G
(R)-3-Phenylpiperidine 97%
(R)-3-Phenylpiperidine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₁H₁₅N
Molecular Weight: 161,244 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03839934
CAS-Nummer: 430461-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR