2-(Methoxycarbonyl)nicotinic acid 96%

BLDPBD150123-250MGEA 37 EUR
BLDPBD150123-250MG BLDPBD150123-100MG BLDPBD150123-5G BLDPBD150123-1G
2-(Methoxycarbonyl)nicotinic acid 96%
2-(Methoxycarbonyl)nicotinic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₇NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00111560
CAS-Nummer: 24195-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR