8-Chinolinsulfonsäure 95%

Synonyme: 8-Quinolinesulphonic acid

BD150121-100G BD150121-500G BD150121-5G BD150121-25G BD150121-10G
BLDPBD150121-100GEA 86 EUR
BLDPBD150121-100G BLDPBD150121-500G BLDPBD150121-5G BLDPBD150121-25G BLDPBD150121-10G
8-Chinolinsulfonsäure 95%
8-Chinolinsulfonsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₇NO₃S
Molecular Weight: 209,23 g/mol
Lagertemperatur: Ambient
MDL: MFCD00024037
CAS-Nummer: 85-48-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR