4-Chloro-5-fluoropyrimidine 95%

BLDPBD149842-25GEA 378 EUR
BLDPBD149842-25G BLDPBD149842-1G BLDPBD149842-5G BLDPBD149842-10G
4-Chloro-5-fluoropyrimidine 95%
4-Chloro-5-fluoropyrimidine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₄H₂ClFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09750157
CAS-Nummer: 347418-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR