4-Chloropyrido[3,2-d]pyrimidine 98%

BD149789-250MG BD149789-5G BD149789-1G
BLDPBD149789-250MGEA 68 EUR
BLDPBD149789-250MG BLDPBD149789-5G BLDPBD149789-1G
4-Chloropyrido[3,2-d]pyrimidine 98%
4-Chloropyrido[3,2-d]pyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₄ClN₃
Molecular Weight: 165,5798 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD10566748
CAS-Nummer: 51674-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR