Triethylene glycol monobenzyl ether 96%

Synonyme: 2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethanol2-(2-(2-(Benzyloxy)ethoxy)ethoxy)ethanol

BLDPBD14977-100GEA 396 EUR
BLDPBD14977-100G BLDPBD14977-25G BLDPBD14977-5G BLDPBD14977-1G
Triethylene glycol monobenzyl ether 96%
Triethylene glycol monobenzyl ether

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₂₀O₄
Molecular Weight: 240,30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02093556
CAS-Nummer: 55489-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR