Sodium pyridine-3-sulphonate 98%

Synonyme: Pyridine-3-sulphonic acid Sodium Salt

BD149760-25G BD149760-5G BD149760-100G
BLDPBD149760-25GEA 71 EUR
BLDPBD149760-25G BLDPBD149760-5G BLDPBD149760-100G
Sodium pyridine-3-sulphonate 98%
Sodium pyridine-3-sulphonate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₅NO₃S
Molecular Weight: 181,14 g/mol
Lagertemperatur: Ambient
MDL: MFCD00065041
CAS-Nummer: 15521-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR