2H-Pyran-3,5(4H,6H)-dione 98%

BD149588-1G BD149588-250MG BD149588-10G BD149588-25G BD149588-5G
BLDPBD149588-1GEA 35 EUR
BLDPBD149588-1G BLDPBD149588-250MG BLDPBD149588-10G BLDPBD149588-25G BLDPBD149588-5G
2H-Pyran-3,5(4H,6H)-dione 98%
2H-Pyran-3,5(4H,6H)-dione
Formel: C₅H₆O₃
Molecular Weight: 114,1 g/mol
MDL: MFCD09032525
CAS-Nummer: 61363-56-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR