2-Methyl-2-propanyl-[2-(4-hydroxyphenyl)ethyl]carbamat 97%

Synonyme: N-Boc-4-hydroxyphenethylamine,tert-Butyl 4-hydroxyphenethylcarbamate,N-(tert-Butoxycarbonyl)-4-hydroxyphenethylamine,N-Boc-tyramine,N-(tert-Butoxycarbonyl)tyramine

BD14958-100MG BD14958-10G BD14958-100G BD14958-1G BD14958-500G BD14958-250MG BD14958-5G BD14958-25G
BLDPBD14958-100MGEA 6 EUR
BLDPBD14958-100MG BLDPBD14958-10G BLDPBD14958-100G BLDPBD14958-1G BLDPBD14958-500G BLDPBD14958-250MG BLDPBD14958-5G BLDPBD14958-25G
2-Methyl-2-propanyl-[2-(4-hydroxyphenyl)ethyl]carbamat 97%
tert-Butyl-4-hydroxyphenethylcarbamate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₃H₁₉NO₃
Molecular Weight: 237,2949 g/mol
Lagertemperatur: Ambient
MDL: MFCD05864730
CAS-Nummer: 64318-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR