p-Tolylacetat 98%

BLDPBD149518-100GEA 34 EUR
BLDPBD149518-100G BLDPBD149518-25G BLDPBD149518-500G
p-Tolylacetat 98%
p-Tolylacetat

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₀O₂
Molecular Weight: 150,1745 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008703
CAS-Nummer: 140-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR