4-Aminobenzanilid 97%

Synonyme: 4-Aminobenzanilid

BLDPBD1495-10GEA 231 EUR
BLDPBD1495-10G BLDPBD1495-100MG BLDPBD1495-1G BLDPBD1495-250MG BLDPBD1495-5G
4-Aminobenzanilid 97%
4-Amino-N-benzanilid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₃H₁₂N₂O
Molecular Weight: 212,2472 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017107
CAS-Nummer: 782-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR