p-Toluenesulphinic acid sodium salt 97%

Synonyme: Sodium 4-methylbenzenesulphinate,Sodium-p-toluenesulphinate,4-Methylbenzenesulphinic acid sodium salt,Sodiumtoluene-4-sulphinate

BD149483-100G BD149483-1000G BD149483-25G BD149483-500G
BLDPBD149483-100GEA 11 EUR
BLDPBD149483-100G BLDPBD149483-1000G BLDPBD149483-25G BLDPBD149483-500G
p-Toluenesulphinic acid sodium salt 97%
p-Toluenesulphinic acid sodium salt

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₇NaO₂S
Molecular Weight: 178,19 g/mol
Lagertemperatur: Ambient
MDL: MFCD00013136
CAS-Nummer: 824-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR