Propylenglycol 99%

Synonyme: Propylenglycol,(±)-1,2-Propylenglycol,(±)-1,2-Dihydroxypropan

BD149321-500G BD149321-1000G BD149321-100G BD149321-25G
BLDPBD149321-500GEA 11 EUR
BLDPBD149321-500G BLDPBD149321-1000G BLDPBD149321-100G BLDPBD149321-25G
Propylenglycol 99%
Propylenglycol

Storage condition: Inert atmosphere, Room Temperature

Formel: CH₃CH(OH)CH₂OH
Molecular Weight: 76,1 g/mol
Lagertemperatur: Ambient
MDL: MFCD00064272
CAS-Nummer: 57-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR