Propylcyclohexan 98%

Synonyme: 1-Cyclohexylpropane

BD149319-1G BD149319-25G BD149319-5G
BLDPBD149319-1GEA 9 EUR
BLDPBD149319-1G BLDPBD149319-25G BLDPBD149319-5G
Propylcyclohexan 98%
Propylcyclohexane

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₈
Molecular Weight: 126,24 g/mol
Lagertemperatur: Ambient
MDL: MFCD00013770
CAS-Nummer: 1678-92-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR