Propyl-3,4,5-trihydroxybenzoat 98%

Synonyme: 3,4,5-Trihydroxybenzoesäurepropylester,Propylgallat,Gallussäurepropylester,Progalline P

BD149297-25G
BLDPBD149297-25GEA 7 EUR
BLDPBD149297-25G
Propyl-3,4,5-trihydroxybenzoat 98%
Propyl-3,4,5-trihydroxybenzoat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₂O₅
Molecular Weight: 212,2 g/mol
Lagertemperatur: Ambient
MDL: MFCD00002196
CAS-Nummer: 121-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR