4-Hexylbenzoesäure 98%

BLDPBD14927-25GEA 224 EUR
BLDPBD14927-25G BLDPBD14927-100G BLDPBD14927-1G BLDPBD14927-5G BLDPBD14927-0.025G
4-Hexylbenzoesäure 98%
4-Hexylbenzoesäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₈O₂
Molecular Weight: 206,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013997
CAS-Nummer: 21643-38-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR