Potassium tripolyphosphate 96%

BLDPBD148944-100GEA 11 EUR
BLDPBD148944-100G BLDPBD148944-500G BLDPBD148944-1000G
Potassium tripolyphosphate 96%
Potassium tripolyphosphate

Storage condition: Inert atmosphere, Room Temperature

Formel: K₅O₁₀P₃
Molecular Weight: 448,41 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00054129
CAS-Nummer: 13845-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR