Kaliumtrifluoracetat 98%

Synonyme: Trifluoressigsäure Kaliumsalz

BD148938-100G BD148938-25G BD148938-10G BD148938-500G BD148938-1000G BD148938-5G
BLDPBD148938-100GEA 22 EUR
BLDPBD148938-100G BLDPBD148938-25G BLDPBD148938-10G BLDPBD148938-500G BLDPBD148938-1000G BLDPBD148938-5G
Kaliumtrifluoracetat 98%
Kaliumtrifluoracetat

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₂F₃KO₂
Molecular Weight: 152,11 g/mol
Lagertemperatur: Ambient
MDL: MFCD00013215
CAS-Nummer: 2923-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR