Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate 98%

BLDPBD148920-5GEA 2390 EUR
BLDPBD148920-5G BLDPBD148920-250MG BLDPBD148920-1G BLDPBD148920-10MG BLDPBD148920-100MG BLDPBD148920-50MG
Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate 98%
Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₃₂H₁₂BF₂₄K
Molecular Weight: 902,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077934
CAS-Nummer: 105560-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR