Potassiumpyruvate 99+%

BD148879-1000G BD148879-25G BD148879-100G BD148879-500G
BLDPBD148879-1000GEA 187 EUR
BLDPBD148879-1000G BLDPBD148879-25G BLDPBD148879-100G BLDPBD148879-500G
Potassiumpyruvate 99+%
Potassiumpyruvate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₃H₃KO₃
Molecular Weight: 126,15 g/mol
Lagertemperatur: Ambient
MDL: MFCD00053313
CAS-Nummer: 4151-33-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR