Kalium-D-gluconat 99%

Synonyme: Dextronsäure KaliumsalzD-Gluconsäure Kaliumsalz(2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexansäure Kaliumsalz

BLDPBD148782-25GEA 6 EUR
BLDPBD148782-0.025 BLDPBD148782-25G BLDPBD148782-100G BLDPBD148782-500G
Kalium-D-gluconat 99%
Kalium-D-gluconat

Storage condition: Inert atmosphere, Room Temperature

Formel: HOH₂C(CH(OH))₄COOK
Molecular Weight: 234,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00064211
CAS-Nummer: 299-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR