Kalium(4-chlorphenyl)trifluoroborat 97%

Synonyme: (4-Chlorphenyl)trifluorborsäure Kaliumsalz

BD148719-250MG BD148719-10G BD148719-1G BD148719-25G BD148719-5G BD148719-100G
BLDPBD148719-250MGEA 8 EUR
BLDPBD148719-250MG BLDPBD148719-10G BLDPBD148719-1G BLDPBD148719-25G BLDPBD148719-5G BLDPBD148719-100G
Kalium(4-chlorphenyl)trifluoroborat 97%
Kalium(4-chlorphenyl)trifluoroborat

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₄BClF₃K
Molecular Weight: 218,45 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD04115770
CAS-Nummer: 661465-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR