Kalium(4-acetylphenyl)trifluoroborat 98%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD148715-25GEA
235
EUR
BLDPBD148715-25G
BLDPBD148715-5G
BLDPBD148715-1G
Kalium(4-acetylphenyl)trifluoroborat 98%
Kalium(4-acetylphenyl)trifluoroborat
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₈H₇BF₃KO Molecular Weight: 226,05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07784376 CAS-Nummer: 252726-24-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...