Kalium(2-formylphenyl)trifluoroborat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD148701-1GEA 59 EUR
BLDPBD148701-1G BLDPBD148701-100MG BLDPBD148701-5G BLDPBD148701-250MG
Kalium(2-formylphenyl)trifluoroborat 98%
Kalium(2-formylphenyl)trifluoroborat

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₅BF₃KO
Molecular Weight: 212,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02093979
CAS-Nummer: 192863-39-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR