Polydatin 99+%

Synonyme: (2S,3R,4S,5S,6R)-2-(3-Hydroxy-5-(4-hydroxystyryl)phenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

BD148564-25G BD148564-100G BD148564-1G BD148564-500G BD148564-5G
BLDPBD148564-25GEA 58 EUR
BLDPBD148564-25G BLDPBD148564-100G BLDPBD148564-1G BLDPBD148564-500G BLDPBD148564-5G
Polydatin 99+%
Polydatin

Storage condition: Sealed in dry, 2-8°C

Formel: C₂₀H₂₂O₈
Molecular Weight: 390,3839 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00210592
CAS-Nummer: 65914-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR