Propynol ethoxylate 90%

BLDPBD148401-5GEA 10 EUR
BLDPBD148401-5G BLDPBD148401-25G
Propynol ethoxylate 90%
Propynol ethoxylate

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₅H₈O₂
Molecular Weight: 100,12 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00099294
CAS-Nummer: 3973-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR