Platin(II)acetylacetonat 98%

Synonyme: Platin(II)-bis(2,4-pentandionato-O,O'),Platin(II)-2,4-pentandionat,Bis(2,4-pentandionato-O,O')platin(II),Di(2,4-pentandionato-O,O')platin(II)

BD148380-100MG BD148380-5G BD148380-250MG BD148380-1G
BLDPBD148380-100MGEA 19 EUR
BLDPBD148380-100MG BLDPBD148380-5G BLDPBD148380-250MG BLDPBD148380-1G
Platin(II)acetylacetonat 98%
Platin(II)acetylacetonat

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₁₄O₄Pt
Molecular Weight: 393,3 g/mol
Lagertemperatur: Ambient
MDL: MFCD00000028
CAS-Nummer: 15170-57-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR