8-Ethyl-5-oxo-2-(pyrrolidin-1-yl)-5,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylic acid 98%

BD148337-100MG BD148337-250MG
BLDPBD148337-100MGEA 308 EUR
BLDPBD148337-100MG BLDPBD148337-250MG
8-Ethyl-5-oxo-2-(pyrrolidin-1-yl)-5,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylic acid 98%
8-Ethyl-5-oxo-2-(pyrrolidin-1-yl)-5,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₄H₁₆N₄O₃
Molecular Weight: 288,30 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00058574
CAS-Nummer: 19562-30-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR