Piroctone olamine 97%

Synonyme: 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridone Ethanolamine,1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)pyridin-2(1H)-one 2-aminoethanol salt

BD148329-25G BD148329-1G BD148329-5G
BLDPBD148329-25GEA 160 EUR
BLDPBD148329-25G BLDPBD148329-1G BLDPBD148329-5G
Piroctone olamine 97%
Piroctone olamine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₄H₂₃NO₂
Molecular Weight: 298,43 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD01690792
CAS-Nummer: 68890-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR