3-Carboxyphenylboronsäurepinakolester 98%

BLDPBD14830-10GEA 31 EUR
BLDPBD14830-10G BLDPBD14830-1G BLDPBD14830-25G BLDPBD14830-5G BLDPBD14830-250MG
3-Carboxyphenylboronsäurepinakolester 98%
3-Carboxyphenylboronsäurepinakolester

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₃H₁₇BO₄
Molecular Weight: 248,083 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03411930
CAS-Nummer: 269409-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR