4-Formylbenzeneboronic acid pinacol ester 96%

Synonyme: 4-Formylphenylboronic acid pinacol ester,4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde,4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

BD14829-100G BD14829-250MG BD14829-25G BD14829-1G BD14829-500G BD14829-5G BD14829-10G
BLDPBD14829-100GEA 270 EUR
BLDPBD14829-100G BLDPBD14829-250MG BLDPBD14829-25G BLDPBD14829-1G BLDPBD14829-500G BLDPBD14829-5G BLDPBD14829-10G
4-Formylbenzeneboronic acid pinacol ester 96%
4-Formylbenzeneboronic acid pinacol ester

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₃H₁₇BO₃
Molecular Weight: 232,086 g/mol
Lagertemperatur: Ambient
MDL: MFCD04972375
CAS-Nummer: 128376-64-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR