3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 98%

Synonyme: 2-(3-Formylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,3-Formylphenylboronic acid, pinacol ester,3-Formylphenylboronic acid Pinacol ester

BD14828-100G BD14828-5G BD14828-250MG BD14828-1G BD14828-10G BD14828-25G
BLDPBD14828-100GEA 336 EUR
BLDPBD14828-100G BLDPBD14828-5G BLDPBD14828-250MG BLDPBD14828-1G BLDPBD14828-10G BLDPBD14828-25G
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 98%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₃H₁₇BO₃
Molecular Weight: 232,09 g/mol
Lagertemperatur: Ambient
MDL: MFCD05663872
CAS-Nummer: 380151-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR