PIPES Dinatriumsalz 98%

Synonyme: 2,2'-(Piperazin-1,4-diyl)bis(ethansulfonsäure) Dinatriumsalz,PIPES-Na2,Piperazin-1,4-bis(2-ethansulfonsäure) Dinatriumsalz,Dinatriumpiperazin-1,4-diethansulfonat,Piperazin-N,N'-bis-2-ethansulfonsäure Dinatriumsalz

BD148265-500G BD148265-1000G BD148265-25G BD148265-100G
BLDPBD148265-500GEA 71 EUR
BLDPBD148265-500G BLDPBD148265-1000G BLDPBD148265-25G BLDPBD148265-100G
PIPES Dinatriumsalz 98%
PIPES Dinatriumsalz

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₁₆N₂Na₂O₆S₂
Molecular Weight: 346,34 g/mol
Lagertemperatur: Ambient
MDL: MFCD00064349
CAS-Nummer: 76836-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR