N,N-Dimethylpiperazine-1-sulphonamide 95%

BLDPBD148174-1GEA 95 EUR
BLDPBD148174-1G BLDPBD148174-100MG BLDPBD148174-250MG
N,N-Dimethylpiperazine-1-sulphonamide 95%
N,N-Dimethylpiperazine-1-sulphonamide

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₆H₁₅N₃O₂S
Molecular Weight: 193,267 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04972523
CAS-Nummer: 98961-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR